Skip to Content
MilliporeSigma

XPIH9BCFAE9D

AiFChem (an XtalPi company)

4-Phenylbutanoic acid

Synonym(s):

4-phenylbutanoic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C10H12O2
CAS Number:
Molecular Weight:
164.2
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C10H12O2/c11-10(12)8-4-7-9-5-2-1-3-6-9/h1-3,5-6H,4,7-8H2,(H,11,12)

Inchi Key

OBKXEAXTFZPCHS-UHFFFAOYSA-N

Purity

99%

Country of Origin

CN

Physical Form

Form: Crystal - Powder

Shipping Temperature

Keep container tightly closed in a dry and well-ventilated area. Containers which are opened must be carefully@resealed and kept upright to prevent leakage.Sealed in dry,Room Temperature

Storage temp.

Keep container tightly closed in a dry and well-ventilated area. Containers which are opened must be carefully@resealed and kept upright to prevent leakage.Sealed in dry,Room Temperature



Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library