Skip to Content
MilliporeSigma

XPIH9BCFED81

AiFChem (an XtalPi company)

4-Bromo-3-nitrobenzaldehyde

Synonym(s):

4-bromo-3-nitrobenzaldehyde

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C7H4O3N1Br1
CAS Number:
Molecular Weight:
230.02
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C7H4BrNO3/c8-6-2-1-5(4-10)3-7(6)9(11)12/h1-4H

Inchi Key

SAFSVELFSYQXOV-UHFFFAOYSA-N

Purity

97%

Country of Origin

CN

Physical Form

Form: Crystal - Powder / Colour: Pale yellow - Pale reddish

Shipping Temperature

Keep container tightly closed in a dry and well-ventilated area. Containers which are opened must be carefully@resealed and kept upright to prevent leakage.Keep in dark place,Sealed in dry,Room Temperature

Storage temp.

Keep container tightly closed in a dry and well-ventilated area. Containers which are opened must be carefully@resealed and kept upright to prevent leakage.Keep in dark place,Sealed in dry,Room Temperature



Still not finding the right product?

Explore all of our products under 4-Bromo-3-nitrobenzaldehyde


signalword

Warning

pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library