Skip to Content
MilliporeSigma

XPIH9BD041EC

AiFChem (an XtalPi company)

(2R,3R,4R,5R)-2,3,4,5,6-Pentahydroxyhexanal

Synonym(s):

D-allose

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C6H12O6
CAS Number:
Molecular Weight:
180.16
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C6H12O6/c7-1-3(9)5(11)6(12)4(10)2-8/h1,3-6,8-12H,2H2/t3-,4+,5-,6+/m0/s1

Inchi Key

GZCGUPFRVQAUEE-BGPJRJDNSA-N

Shipping Temperature

Keep container tightly closed in a dry and well-ventilated area. Containers which are opened must be carefully@resealed and kept upright to prevent leakage.Inert atmosphere,2-8C

Purity

98%

Storage temp.

Keep container tightly closed in a dry and well-ventilated area. Containers which are opened must be carefully@resealed and kept upright to prevent leakage.Inert atmosphere,2-8C

Country of Origin

CN

Physical Form

White to almost white powder to crystal

Boiling Point

No data available

Melting Point

No data available

Flash Point

No data available



Still not finding the right product?


signalword

Warning

pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library