Skip to Content
MilliporeSigma

XPIH9BD0691C

AiFChem (an XtalPi company)

3,6-Bis(dimethylamino)xanthylium chloride

Synonym(s):

3,6-bis(dimethylamino)xanthenium chloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C17H19O1N2Cl1
CAS Number:
Molecular Weight:
302.8
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C17H19N2O.ClH/c1-18(2)14-7-5-12-9-13-6-8-15(19(3)4)11-17(13)20-16(12)10-14;/h5-11H,1-4H3;1H/q+1;/p-1

Inchi Key

INCIMLINXXICKS-UHFFFAOYSA-M

Purity

55%

Country of Origin

CN

Physical Form

Solid

Shipping Temperature

Keep container tightly closed in a dry and well-ventilated area. Containers which are opened must be carefully@resealed and kept upright to prevent leakage.Sealed in dry,Room Temperature

Storage temp.

Keep container tightly closed in a dry and well-ventilated area. Containers which are opened must be carefully@resealed and kept upright to prevent leakage.Sealed in dry,Room Temperature



Still not finding the right product?


signalword

Danger

pictograms

Skull and crossbonesHealth hazard


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library