Storage temp.
Room temperature
Inchi Code
1S/C12H18O2/c13-11(14)7-12-4-8-1-9(5-12)3-10(2-8)6-12/h8-10H,1-7H2,(H,13,14)
Inchi Key
AOTQGWFNFTVXNQ-UHFFFAOYSA-N
Shipping Temperature
Normal
Purity
98%
Country of Origin
US
Physical Form
Solid
Melting Point
134-137C
Density
1.167
Still not finding the right product?
Explore all of our products under 1-Adamantaneacetic acid
hstatements
Signal Word
Warning
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
