Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ALNH9A9D6425

Aladdin Scientific

Ammonium phosphatedi basic

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
132.06
MDL number:

mp

155 C

density

1.619

Iupac Name

diammonium hydrogen phosphate

Inchi Code

1S/2H3N.H3O4P/c;;1-5(2,3)4/h2*1H3;(H3,1,2,3,4)

InChI key

MNNHAPBLZZVQHP-UHFFFAOYSA-N

Purity

98.5%

Country of Origin

US

Shipping Temperature

Normal

Product Link

https://www.aladdinsci.com/ammonium-phosphatedi-basic-bioactive-small-molecules-by-aladdin-scientific-a112553.html

Melting Point

155 C

density

1.619

Hazard Statements

Precautionary statements

MSDS

https://ald-pub-files.oss-cn-shanghai.aliyuncs.com/aladdinsci/pdp/sds/1/A112553-SCI_1bd5dfb4f10793a799e6a24be93e7a3b.pdf


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service