Skip to Content
MilliporeSigma

ALNH9A9E6664

Aladdin Scientific

2,3-Dihydrofuran

Synonym(s):

2,3-dihydrofuran

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C4H6O1
CAS Number:
Molecular Weight:
70.09
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C4H6O/c1-2-4-5-3-1/h1,3H,2,4H2

Inchi Key

JKTCBAGSMQIFNL-UHFFFAOYSA-N

Shipping Temperature

Wet ice

Purity

99%

Storage temp.

Store at 2-8C;Argon charged

Country of Origin

US

Physical Form

Liquid

Boiling Point

54-55C

Flash Point

-24C

Density

0.93



Still not finding the right product?

Explore all of our products under 2,3-Dihydrofuran



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library