Skip to Content
MilliporeSigma

ALNH9A9E66A4

Aladdin Scientific

tert-Dodecylmercaptan (mixture of isomers)

Sign Into View Organizational & Contract Pricing

Select a Size


About This Item

CAS Number:
Molecular Weight:
202.4
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

mp

227-248C

Iupac Name

2,3,3,4,4,5-hexamethylhexane-2-thiol

Inchi Code

1S/C12H26S/c1-9(2)10(3,4)11(5,6)12(7,8)13/h9,13H,1-8H3

InChI key

YAJYJWXEWKRTPO-UHFFFAOYSA-N

Purity

98%

Country of Origin

US

Shipping Temperature

Normal

Product Link

https://www.aladdinsci.com/tert-dodecylmercaptan-mixture-of-isomers-bioactive-small-molecules-by-aladdin-scientific-d106952.html

Flash Point

97 C

Melting Point

227-248C

pictograms

EnvironmentExclamation mark

Signal Word

Warning

Hazard Statements

MSDS

https://ald-pub-files.oss-cn-shanghai.aliyuncs.com/aladdinsci/pdp/sds/1/D106952-SCI_73728bc52e5827869c2f1a4fc7dc9470.pdf


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service