Skip to Content
MilliporeSigma

ALNH9A9E68A0

Aladdin Scientific

Dodecylamine

Synonym(s):

1-dodecanamine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C12H27N1
CAS Number:
Molecular Weight:
185.35
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C12H27N/c1-2-3-4-5-6-7-8-9-10-11-12-13/h2-13H2,1H3

Inchi Key

JRBPAEWTRLWTQC-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Room temperature;Argon charged

Country of Origin

US

Boiling Point

247-249 C

Melting Point

26 C

Flash Point

115C

Density

0.806



Still not finding the right product?

Explore all of our products under Dodecylamine



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library