Skip to Content
MilliporeSigma

ALNH9A9E69C8

Aladdin Scientific

3,5-Dichloropyridine

Synonym(s):

3,5-dichloropyridine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C5H3N1Cl2
CAS Number:
Molecular Weight:
147.99
MDL number:


Inchi Code

1S/C5H3Cl2N/c6-4-1-5(7)3-8-2-4/h1-3H

Inchi Key

WPGHPGAUFIJVJF-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Country of Origin

US

Physical Form

Solid

Boiling Point

178C

Melting Point

65C



Still not finding the right product?

Explore all of our products under 3,5-Dichloropyridine



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library