Inchi Code
1S/C18H13N5O6/c24-21(25)15-11-16(22(26)27)18(17(12-15)23(28)29)19-20(13-7-3-1-4-8-13)14-9-5-2-6-10-14/h1-12,19H
Inchi Key
WCBPJVKVIMMEQC-UHFFFAOYSA-N
Shipping Temperature
Normal
Purity
98%
Country of Origin
US
Melting Point
~174C(dec.)(lit.)
hstatements
signalword
Danger
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.


