Skip to Content
MilliporeSigma

ALNH9A9F520A

Aladdin Scientific

1-Methylpyrrole

Synonym(s):

1-methyl-1H-pyrrole

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
Molecular Weight:
81.12
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


bp

111-114C

mp

-57C

Inchi Code

1S/C5H7N/c1-6-4-2-3-5-6/h2-5H,1H3

Inchi Key

OXHNLMTVIGZXSG-UHFFFAOYSA-N

Storage temp.

Argon charged

Purity

99%

Country of Origin

US

Shipping Temperature

Normal

Flash Point

16 C



MSDS

https://ald-pub-files.oss-cn-shanghai.aliyuncs.com/aladdinsci/pdp/sds/1/M100701-SCI_2b9bb23c87bbe6f660076cdbf954cbda.pdf

hstatements

signalword

Danger

pictograms

Exclamation markFlame


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library