Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ALNH9A9F542B

Aladdin Scientific

Memantine Hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
215.77
MDL number:

mp

292C

density

0.803

Iupac Name

3,5-dimethyl-1-adamantanamine hydrochloride

Inchi Code

1S/C12H21N.ClH/c1-10-3-9-4-11(2,6-10)8-12(13,5-9)7-10;/h9H,3-8,13H2,1-2H3;1H

InChI key

LDDHMLJTFXJGPI-UHFFFAOYSA-N

Purity

98%

Country of Origin

US

Shipping Temperature

Normal

Product Link

https://www.aladdinsci.com/memantine-hydrochloride-bioactive-small-molecules-by-aladdin-scientific-m107928.html

Melting Point

292C

density

0.803

pictograms

Skull and crossbonesExclamation mark

Signal Word

Danger

MSDS

https://ald-pub-files.oss-cn-shanghai.aliyuncs.com/aladdinsci/pdp/sds/1/M107928-SCI_d8f37a21af4a0ba1ed0365490183b16f.pdf


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service