Skip to Content
MilliporeSigma

ALNH9A9F77AC

Aladdin Scientific

2-Methylpyrrolidine

Synonym(s):

2-methylpyrrolidine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View



bp

97-99 C (lit.) at 744 mmHg

mp

-90 C

Inchi Code

1S/C5H11N/c1-5-3-2-4-6-5/h5-6H,2-4H2,1H3

Inchi Key

RGHPCLZJAFCTIK-UHFFFAOYSA-N

Storage temp.

Store at 2-8C

Purity

97%

Country of Origin

US

Shipping Temperature

Wet ice

Flash Point

2C



Still not finding the right product?

Explore all of our products under 2-Methylpyrrolidine


MSDS

https://ald-pub-files.oss-cn-shanghai.aliyuncs.com/aladdinsci/pdp/sds/1/M341863-SCI_ec35f7cea67df2edd0ffebc703457abf.pdf

hstatements

signalword

Danger



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library