Skip to Content
MilliporeSigma

ALNH9A9FB3C0

Aladdin Scientific

2-Naphthoic acid

Synonym(s):

2-naphthoic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C11H8O2
CAS Number:
Molecular Weight:
172.18
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Storage temp.

Room temperature

Inchi Code

1S/C11H8O2/c12-11(13)10-6-5-8-3-1-2-4-9(8)7-10/h1-7H,(H,12,13)

Inchi Key

UOBYKYZJUGYBDK-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Country of Origin

US

Physical Form

Solid

Melting Point

185-187 C

Flash Point

205C

Density

1.08



Still not finding the right product?

Explore all of our products under 2-Naphthoic acid


signalword

Warning

pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library