MilliporeSigma
All Photos(1)

247677

Sigma-Aldrich

Sodium borohydride

90% NaBH4 basis, pellets, diam. 8 mm, Cobalt (Doped)

Sign Into View Organizational & Contract Pricing

Select a Size

Synonym(s):
Cobalt-doped sodium borohydride, Sodium tetrahydridoborate
Linear Formula:
NaBH4
CAS Number:
Molecular Weight:
37.83
MDL number:
PubChem Substance ID:

Quality Level

Assay

90% NaBH4 basis

form

pellets

composition

Cobalt (Doped)

reaction suitability

reagent type: reductant

diam.

8 mm

impurities

7.5% CoCl2

mp

>300 °C (dec.) (lit.)

SMILES string

[BH4-].[Na+]

InChI

1S/BH4.Na/h1H4;/q-1;+1

InChI key

YOQDYZUWIQVZSF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Sodium borohydride has been used as a reductant in the synthesis of branched gold nanoparticles from gold(III) chloride trihydrate. It has also been used for the production of hydrogen gas by reacting with sea water in the presence of boron oxide.

Features and Benefits

A convenient solid source of hydrogen gas. Each pellet readily liberates ~500mL of hydrogen gas upon contact with water. Useful for producing an anaerobic reducing atmosphere in biological experiments. Cobalt-doped

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.

Visit the Document Library

Difficulty Finding Your Product Or Lot/Batch Number?

Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (example: T1503).

Example:

T1503
Product Number
-
25G
Pack Size/Quantity

Additional examples:

705578-5MG-PW

PL860-CGA/SHF-1EA

MMYOMAG-74K-13

1000309185

enter as 1.000309185)

Having trouble? Feel free to contact Technical Service for assistance.

Lot and Batch Numbers can be found on a product's label following the words 'Lot' or 'Batch'.

Aldrich Products

  • For a lot number such as TO09019TO, enter it as 09019TO (without the first two letters 'TO').

  • For a lot number with a filling-code such as 05427ES-021, enter it as 05427ES (without the filling-code '-021').

  • For a lot number with a filling-code such as STBB0728K9, enter it as STBB0728 without the filling-code 'K9'.

Not Finding What You Are Looking For?

In some cases, a COA may not be available online. If your search was unable to find the COA you can request one.

Request COA

Customers Also Viewed

Slide 1 of 3

1 of 3

Sodium borohydride fine granular for synthesis

Sigma-Aldrich

8.06373

Sodium borohydride

Hydrochloric acid ACS reagent, 37%

Sigma-Aldrich

320331

Hydrochloric acid

Fatemeh Haji Ghahremani et al.
International journal of hyperthermia : the official journal of European Society for Hyperthermic Oncology, North American Hyperthermia Group, 27(6), 625-636 (2011-08-19)
Hyperthermia, generated using microwave (MW), infrared, ultrasound and other methods, is often utilised as an adjuvant technique to sensitise cancer lesions to radiation therapy and chemotherapy. In the current research, MW hyperthermia efficacy in synergism with chemotherapy was investigated in
Acid acceleration of hydrogen generation using seawater as a reactant
Becker-Glad CA & Glad W. E
International Journal of Hydrogen Energy, 41(40), 17761-17770 (2016)
Comparative study of X-ray treatment and photodynamic therapy by using 5-aminolevulinic acid conjugated gold nanoparticles in a melanoma cell line
Mohammadi Z, et al
Artificial Cells, Nanomedicine, and Biotechnology (Print), 45(3), 467-473 (2017)
Dimethylsulphoxide (DMSO) in biological samples: A comparison of the TiCl3 and NaBH4 reduction methods using headspace analysis.
Deschaseaux ESM, et al.
Marine Chemistry, 164, 9-15 (2014)
Enhancement of Cisplatin Efficacy by Gold Nanoparticles or Microwave Hyperthermia? An In Vitro Study on a Melanoma Cell Line
Moradpoor R, et al
Iranian journal of cancer prevention, 10(1) (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service