Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

589187

Sigma-Aldrich

Isophthaloyl-2,2′-13C2 chloride

99 atom % 13C

Synonym(s):

1,3-Benzenedi(carbonyl-13C) dichloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13C2C6H4Cl2O2
CAS Number:
Molecular Weight:
205.01
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:

isotopic purity

99 atom % 13C

form

solid

mp

43-44 °C (lit.)

mass shift

M+2

SMILES string

Cl[13C](=O)c1cccc(c1)[13C](Cl)=O

InChI

1S/C8H4Cl2O2/c9-7(11)5-2-1-3-6(4-5)8(10)12/h1-4H/i7+1,8+1

InChI key

FDQSRULYDNDXQB-BFGUONQLSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

pictograms

CorrosionExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

target_organs

Respiratory system

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service