Skip to Content
MilliporeSigma
Sign Into View Organizational & Contract Pricing

About This Item

Linear Formula:
134Xe
CAS Number:
Molecular Weight:
133.91
MDL number:
UNSPSC Code:
12142001
PubChem Substance ID:

isotopic purity

60 atom %

assay

≥99.5% (Total Xenon)

form

gas

impurities

<0.5% Total others

mass shift

M+2

SMILES string

[134Xe]

InChI

1S/Xe/i1+3

InChI key

FHNFHKCVQCLJFQ-AKLPVKDBSA-N

Looking for similar products? Visit Product Comparison Guide

Application

- Quality control reference standards
- Analytical method development
- Environmental monitoring applications
- Pharmaceutical formulation development

Features and Benefits

Features
  • Improves the quality of NMR data.
  • Enhances understanding of molecular interactions.
  • Facilitates the discoveryof new pharmaceuticals.
  • Supports high-quality outcomes in research projects.


Benefits
  • Enhances the accuracy of metabolic studies.
  • Increases efficiency in drug development processes.
  • Supports regulatory compliance and safety standards.
  • Provides a competitive advantage in the pharmaceutical market.

Packaging

All gas volumes are quoted at Standard Temperature and Pressure (STP) and not at Normal Temperature and Pressure (NTP). One STP Liter is 7.7% more gas than one NTP Liter.
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

pictograms

Gas cylinder

signalword

Warning

hcodes

Hazard Classifications

Press. Gas Compr. Gas

Storage Class

2A - Gases

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service