MilliporeSigma
All Photos(1)

Documents

720100

Sigma-Aldrich

Potassium palmitate-16-d

98 atom % D, 97% (CP)

Sign Into View Organizational & Contract Pricing

Synonym(s):
Hexadecanoic acid-16-d potassium salt, Palmitic acid-16-d potassium salt, Potassium hexadecanoate-16-d
Linear Formula:
CH2D(CH2)14CO2K
CAS Number:
Molecular Weight:
295.52
MDL number:
PubChem Substance ID:

isotopic purity

98 atom % D

assay

97% (CP)

form

solid

mp

285 °C (dec.)

mass shift

M+1

SMILES string

[K+].[2H]CCCCCCCCCCCCCCCC([O-])=O

InChI

1S/C16H32O2.K/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16(17)18;/h2-15H2,1H3,(H,17,18);/q;+1/p-1/i1D;

InChI key

MQOCIYICOGDBSG-RWQOXAPSSA-M

Related Categories

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
605751676454614378
Potassium palmitate-16-d 98 atom % D, 97% (CP)

720100

Potassium palmitate-16-d

Potassium palmitate-13C16 99 atom % 13C, 99% (CP)

605751

Potassium palmitate-13C16

Potassium palmitate-1,3,5,7,9-13C5 endotoxin tested, 99 atom % 13C, 98% (CP)

676454

Potassium palmitate-1,3,5,7,9-13C5

Potassium palmitate-d31 98 atom % D

614378

Potassium palmitate-d31

assay

97% (CP)

assay

99% (CP)

assay

98% (CP)

assay

-

mass shift

M+1

mass shift

M+16

mass shift

M+5

mass shift

M+31

form

solid

form

solid

form

solid

form

solid

mp

285 °C (dec.)

mp

285 °C (lit.)

mp

285 °C (dec.)

mp

285 °C (dec.) (lit.)

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.

Visit the Document Library

Difficulty Finding Your Product Or Lot/Batch Number?

Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (example: T1503).

Example:

T1503
Product Number
-
25G
Pack Size/Quantity

Additional examples:

705578-5MG-PW

PL860-CGA/SHF-1EA

MMYOMAG-74K-13

1000309185

enter as 1.000309185)

Having trouble? Feel free to contact Technical Service for assistance.

Lot and Batch Numbers can be found on a product's label following the words 'Lot' or 'Batch'.

Aldrich Products

  • For a lot number such as TO09019TO, enter it as 09019TO (without the first two letters 'TO').

  • For a lot number with a filling-code such as 05427ES-021, enter it as 05427ES (without the filling-code '-021').

  • For a lot number with a filling-code such as STBB0728K9, enter it as STBB0728 without the filling-code 'K9'.

Not Finding What You Are Looking For?

In some cases, a COA may not be available online. If your search was unable to find the COA you can request one.

Request COA

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service