MilliporeSigma
All Photos(1)

769274

Sigma-Aldrich

Zirconium(IV) oxide

pellets, diam. × thickness 10 mm × 3.5 mm

Sign Into View Organizational & Contract Pricing

Select a Size

Synonym(s):
Zirconia
Linear Formula:
ZrO2
CAS Number:
Molecular Weight:
123.22
EC Number:
MDL number:
NACRES:
NA.23

form

pellets

diam. × thickness

10 mm × 3.5 mm

impurities

≤0.1% PBB (polybrominated biphenyls)
≤0.1% PBDE (polybrominated diphenyl ethers)

bp

5000 °C (lit.)

mp

2700 °C (lit.)

density

5.89 g/mL at 25 °C (lit.)

cation traces

Cd: ≤0.01%
Co: ≤0.0005%
Cr: ≤0.002%
Cu: ≤0.001%
Fe: ≤0.01%
Hg: ≤0.1%
Pb: ≤0.1%
Ti: ≤0.2%
V: ≤0.005%

SMILES string

O=[Zr]=O

InChI

1S/2O.Zr

Inchi Key

MCMNRKCIXSYSNV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Zirconium Dioxide, ZrO2, is a high refractive index, low absorption material used for optical coatings in the near-UV to IR spectral regions.

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.

Visit the Document Library

Difficulty Finding Your Product Or Lot/Batch Number?

Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (example: T1503).

Example:

T1503
Product Number
-
25G
Pack Size/Quantity

Additional examples:

705578-5MG-PW

PL860-CGA/SHF-1EA

MMYOMAG-74K-13

1000309185

enter as 1.000309185)

Having trouble? Feel free to contact Technical Service for assistance.

Lot and Batch Numbers can be found on a product's label following the words 'Lot' or 'Batch'.

Aldrich Products

  • For a lot number such as TO09019TO, enter it as 09019TO (without the first two letters 'TO').

  • For a lot number with a filling-code such as 05427ES-021, enter it as 05427ES (without the filling-code '-021').

  • For a lot number with a filling-code such as STBB0728K9, enter it as STBB0728 without the filling-code 'K9'.

Not Finding What You Are Looking For?

In some cases, a COA may not be available online. If your search was unable to find the COA you can request one.

Request COA

A high-performance spectrally-selective solar absorber based on a yttria-stabilized zirconia cermet with high-temperature stability
Cao, F., et al.
Energy & Environmental Science, 8, 3040-3048 (2015)
Silica?zirconia alkali-resistant coatings by sol?gel route
Dhere, SL.
Current Science, 108(9), 1647-1652 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service