Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
13C3C5H14Cl15N3N2
Molecular Weight:
221.64
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.12
Isotopic purity:
≥98 atom %
Assay:
≥98% (CP)
Mass shift:
M+6
Form:
powder
isotopic purity
≥98 atom %
Quality Segment
assay
≥98% (CP)
form
powder
technique(s)
mass spectrometry (MS): suitable
mass shift
M+6
storage temp.
−20°C
SMILES string
CCN[13C]1=[15N][13C](NC(C)C)=[15N][13C](Cl)=[15N]1
InChI
1S/C8H14ClN5/c1-4-10-7-12-6(9)13-8(14-7)11-5(2)3/h5H,4H2,1-3H3,(H2,10,11,12,13,14)/i6+1,7+1,8+1,12+1,13+1,14+1
InChI key
MXWJVTOOROXGIU-ZNLDTMSVSA-N
signalword
Warning
hcodes
Hazard Classifications
Aquatic Acute 1 - Aquatic Chronic 1 - Skin Sens. 1 - STOT RE 2
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.


