MilliporeSigma
All Photos(1)

Documents

900382

Sigma-Aldrich

Adenosine-15N5 5′-monophosphate disodium salt solution

100 mM (in 5mM Tris HCl / H2O), ≥98 atom % 15N, ≥95% (CP)

Sign Into View Organizational & Contract Pricing

Synonym(s):
AMP-15N5 disodium salt solution
Empirical Formula (Hill Notation):
C10H1215N5Na2O7P
Molecular Weight:
396.15
NACRES:
NA.12

isotopic purity

≥98 atom % 15N

Quality Level

assay

≥95% (CP)

form

liquid

concentration

100 mM (in 5mM Tris HCl / H2O)

technique(s)

bio NMR: suitable

mass shift

M+5

storage temp.

−20°C

SMILES string

[15NH2]C1=[15N]C=[15N]C2=C1[15N]=C[15N]2[C@@]3([H])[C@@](O)([H])[C@](O)([H])[C@](COP(O[Na])(O[Na])=O)([H])O3

Related Categories

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
650676707783645702
isotopic purity

≥98 atom % 15N

isotopic purity

≥98 atom %

isotopic purity

≥98 atom % 15N

isotopic purity

≥98 atom %

mass shift

M+5

mass shift

M+15

mass shift

M+5

mass shift

M+15

assay

≥95% (CP)

assay

≥95% (CP)

assay

≥95% (CP)

assay

≥95% (CP)

form

liquid

form

-

form

liquid

form

-

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.

Visit the Document Library

Difficulty Finding Your Product Or Lot/Batch Number?

Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (example: T1503).

Example:

T1503
Product Number
-
25G
Pack Size/Quantity

Additional examples:

705578-5MG-PW

PL860-CGA/SHF-1EA

MMYOMAG-74K-13

1000309185

enter as 1.000309185)

Having trouble? Feel free to contact Technical Service for assistance.

Lot and Batch Numbers can be found on a product's label following the words 'Lot' or 'Batch'.

Aldrich Products

  • For a lot number such as TO09019TO, enter it as 09019TO (without the first two letters 'TO').

  • For a lot number with a filling-code such as 05427ES-021, enter it as 05427ES (without the filling-code '-021').

  • For a lot number with a filling-code such as STBB0728K9, enter it as STBB0728 without the filling-code 'K9'.

Not Finding What You Are Looking For?

In some cases, a COA may not be available online. If your search was unable to find the COA you can request one.

Request COA

Eran Mick et al.
eLife, 9 (2020-05-29)
Mitochondrial dysfunction is associated with activation of the integrated stress response (ISR) but the underlying triggers remain unclear. We systematically combined acute mitochondrial inhibitors with genetic tools for compartment-specific NADH oxidation to trace mechanisms linking different forms of mitochondrial dysfunction
Andrew G Manford et al.
Cell, 183(1), 46-61 (2020-09-18)
Metazoan organisms rely on conserved stress response pathways to alleviate adverse conditions and preserve cellular integrity. Stress responses are particularly important in stem cells that provide lifetime support for tissue formation and repair, but how these protective systems are integrated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service