MilliporeSigma
All Photos(1)

Documents

900961

Sigma-Aldrich

P(NDI2OD-T2)

Sign Into View Organizational & Contract Pricing

Synonym(s):
N2200, PNDI-2T, Poly{[N,N′-bis(2-octyldodecyl)-naphthalene-1,4,5,8-bis(dicarboximide)-2,6-diyl]-alt-5,5′-(2,2′-bithiophene)}
Linear Formula:
(C62H88N2O4S2)n
CAS Number:

description

Bandgap: 1.59 eV

Quality Level

form

solid

mol wt

average Mn 50-100 kDa

color

blue to dark blue

mp

308 °C (DSC transition temperature)

solubility

organic solvents: soluble (Chloroform, dichlorobenzene, toluene, xylene, anisole)

λmax

700 nm (optical)

orbital energy

HOMO -5.61 eV 
LUMO -4.02 eV 

PDI

1.5‑3.5

Related Categories

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
700665693014681539
vibrant-m

900961

P(NDI2OD-T2)

vibrant-m

693014

(S)-H8-BINAP

mp

308 °C (DSC transition temperature)

mp

390-400 °C

mp

-

mp

215-219 °C

description

Bandgap: 1.59 eV

description

-

description

-

description

-

mol wt

average Mn 50-100 kDa

mol wt

-

mol wt

-

mol wt

-

color

blue to dark blue

color

-

color

-

color

-

solubility

organic solvents: soluble (Chloroform, dichlorobenzene, toluene, xylene, anisole)

solubility

-

solubility

-

solubility

-

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.

Visit the Document Library

Difficulty Finding Your Product Or Lot/Batch Number?

Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (example: T1503).

Example:

T1503
Product Number
-
25G
Pack Size/Quantity

Additional examples:

705578-5MG-PW

PL860-CGA/SHF-1EA

MMYOMAG-74K-13

1000309185

enter as 1.000309185)

Having trouble? Feel free to contact Technical Service for assistance.

Lot and Batch Numbers can be found on a product's label following the words 'Lot' or 'Batch'.

Aldrich Products

  • For a lot number such as TO09019TO, enter it as 09019TO (without the first two letters 'TO').

  • For a lot number with a filling-code such as 05427ES-021, enter it as 05427ES (without the filling-code '-021').

  • For a lot number with a filling-code such as STBB0728K9, enter it as STBB0728 without the filling-code 'K9'.

Not Finding What You Are Looking For?

In some cases, a COA may not be available online. If your search was unable to find the COA you can request one.

Request COA

n-Channel Semiconductor Materials Design for Organic Complementary Circuits.
Usta H, et al.
Accounts of Chemical Research, 44(7), 501-510 (2011)
Hyunbum Kang et al.
Journal of the American Chemical Society, 137(6), 2359-2365 (2015-01-22)
The molecular weight of a conjugated polymer is one of the key factors determining the electrical, morphological, and mechanical properties as well as its solubility in organic solvents and miscibility with other polymers. In this study, a series of semicrystalline
He Yan et al.
Nature, 457(7230), 679-686 (2009-01-23)
Printed electronics is a revolutionary technology aimed at unconventional electronic device manufacture on plastic foils, and will probably rely on polymeric semiconductors for organic thin-film transistor (OTFT) fabrication. In addition to having excellent charge-transport characteristics in ambient conditions, such materials

Articles

Professor Chen (Nankai University, China) and his team explain the strategies behind their recent record-breaking organic solar cells, reaching a power conversion efficiency of 17.3%.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service