Select a Size
About This Item
type
(High purity Salts)
Quality Level
assay
≥99.9% trace metals basis, 98.0-102.0% (EDTA, complexometric)
form
powder or crystals
greener alternative product characteristics
Catalysis
Learn more about the Principles of Green Chemistry.
sustainability
Greener Alternative Product
impurities
≤1000 ppm (trace metals analysis)
color
faint blue to dark blue-green
mp
56 °C (lit.)
solubility
water: soluble
density
2.05 g/mL at 25 °C (lit.)
anion traces
chloride (Cl-): ≤20 ppm, sulfate (SO42-): ≤50 ppm
cation traces
Al: <100 ppm, Cu: <20 ppm, K: <20 ppm
application(s)
battery manufacturing
greener alternative category
SMILES string
O.O.O.O.O.O.[Ni++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
InChI
1S/2NO3.Ni.6H2O/c2*2-1(3)4;;;;;;;/h;;;6*1H2/q2*-1;+2;;;;;;
InChI key
AOPCKOPZYFFEDA-UHFFFAOYSA-N
General description
Application
Nickel(II) nitrate hexahydrate can be used as
- As a key component in the development of mesoporous NiCo2O4 nanosheets as electrode materials for Li-ion batteries and supercapacitors via microwave method. The NiCo2O4 electrodes exhibited a high reversible capacity , good rate capability and stable cycling performance. In addition, the electrode performs superior pseudocapacitive performance and excellent cycling stability in supercapacitors
- To the synthesis of NiCoSe2@carbon anode for high-performance sodium-ion batteries. The NiCoSe2@carbon exhibited three-dimensional porous carbon structure demonstrating exceptional electrochemical performance as anode materials in sodium-ion batteries with high specific capacity and superior capacity retention rate
- To the synthesis of electrocatalysts like hollow NiCo2O4 microcuboids via hydrothermal method. The hollow NiCo2O4 microcuboids ehibits water splitting including oxygen evolution reaction (OER) and hydrogen evolution reaction (HER). The superior water splitting activity is due to the structural features of the electrode
- As a main component to synthesis 3D hierarchical nickel–iron–cobalt LDH nanosheets/carbon fibers (NiFeCo-LDH/CF) through solvent-thermal treatment of ZIF-67/CF. The modified electronic structure improve the OER activity for the NiFeCo-LDH catalyst
- In addition, it has been used to synthesis Ce dopped NiFe-layered double hydroxide (LDH) electrocatalyst directly on a nickel foam (NF) substrate at room temperature using an electrodeposition technique. It is used as a bifunctional catalyst for water splitting with excellent stability up to 40 hours.The enhanced electrochemical performance of the Ce-doped NiFe-LDH electrocatalyst can be attributed to its unique 3D network structure. This structure increases the surface area available for electrochemical reactions, thus enhancing its overall efficiency. Additionally, the doping of cerium (Ce) generates a greater number of active sites within the catalyst, further augmenting its catalytic capabilities. These combined factors contribute to the catalyst′s improved performance in water splitting applications
Features and Benefits
- Water soluble
- Medium purity (99.9%)
- Low trace metals in ppm level
- Cost effective
- Suitable for Energy applications
Still not finding the right product?
Explore all of our products under Nickel(II) nitrate hexahydrate
signalword
Danger
Hazard Classifications
Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1A - Eye Dam. 1 - Muta. 2 - Ox. Sol. 2 - Repr. 1B - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 1 Inhalation
Storage Class
5.1B - Oxidizing hazardous materials
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
A comprehensive guide to high-purity salt precursors for cathode active materials, advancing lithium-ion battery efficiency, safety, and cycle life.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| 939323-1000G | 04065271973046 |
| 939323-250G | 04065271973053 |




