About This Item
UNSPSC Code:
26142300
NACRES:
NB.35
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
packaging
pack of 1 ea
manufacturer/tradename
Bel-Art F24980-0001
W × L × H
5.2 cm (2 in.) × 10.5 cm (4 in.) × 6.2 cm (2.5 in.)
General description
The top permits enough room for the flip tops on micro tubes. The compact size saves bench top space and is ideal for DNA sequencing.
  • Firm, transparent, acrylic block has a snug top prevents leaking beta radiation
  • Holds thirteen 1.5 mL micro sample tubes or similar with 11 mm diameter
  • Can be use to transfer hazardous samples
Choose from one of the most recent versions:
Certificates of Analysis (COA)