Skip to Content
MilliporeSigma

GF62597986

Copper - O.F.H.C.

tube, 100mm, outside diameter 10mm, inside diameter 8mm, wall thickness 1.0mm, 99.95+%

Synonym(s):

Copper, CV007800

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
Cu
CAS Number:
Molecular Weight:
63.55
NACRES:
NA.23
PubChem Substance ID:
UNSPSC Code:
12141711
MDL number:
eCl@ss:
38150101
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

99.95%

form

tubes

manufacturer/tradename

Goodfellow 625-979-86

resistivity

1.673 μΩ-cm, 20°C

L × wall thickness

100 mm × 1.0 mm

O.D. × I.D.

10 mm × 8 mm

bp

2567 °C (lit.)

mp

1083.4 °C (lit.)

density

8.94 g/mL at 25 °C (lit.)

SMILES string

[Cu]

InChI

1S/Cu

InChI key

RYGMFSIKBFXOCR-UHFFFAOYSA-N

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library