MilliporeSigma
All Photos(2)

Documents

GF65804238

Samarium

foil, 25x25mm, thickness 0.1mm, as rolled, 99%

Sign Into View Organizational & Contract Pricing

Synonym(s):
Samarium, SM000210
Empirical Formula (Hill Notation):
Sm
CAS Number:
Molecular Weight:
150.36
MDL number:
PubChem Substance ID:
NACRES:
NA.23

assay

99%

form

foil

manufacturer/tradename

Goodfellow 658-042-38

resistivity

91.4 μΩ-cm, 0°C

size × thickness

25x25 mm × 0.1 mm

bp

1794 °C (lit.)

mp

1074 °C (lit.)

density

7.47 g/mL at 25 °C (lit.)

SMILES string

[Sm]

InChI

1S/Sm

InChI key

KZUNJOHGWZRPMI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

pictograms

Flame

signalword

Warning

hcodes

Hazard Classifications

Water-react 3

Storage Class

4.3 - Hazardous materials, which set free flammable gases upon contact with water

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.

Visit the Document Library

Difficulty Finding Your Product Or Lot/Batch Number?

Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (example: T1503).

Example:

T1503
Product Number
-
25G
Pack Size/Quantity

Additional examples:

705578-5MG-PW

PL860-CGA/SHF-1EA

MMYOMAG-74K-13

1000309185

enter as 1.000309185)

Having trouble? Feel free to contact Technical Service for assistance.

Lot and Batch Numbers can be found on a product's label following the words 'Lot' or 'Batch'.

Aldrich Products

  • For a lot number such as TO09019TO, enter it as 09019TO (without the first two letters 'TO').

  • For a lot number with a filling-code such as 05427ES-021, enter it as 05427ES (without the filling-code '-021').

  • For a lot number with a filling-code such as STBB0728K9, enter it as STBB0728 without the filling-code 'K9'.

Not Finding What You Are Looking For?

In some cases, a COA may not be available online. If your search was unable to find the COA you can request one.

Request COA

Y Kamochi et al.
Yakugaku zasshi : Journal of the Pharmaceutical Society of Japan, 120(3), 245-255 (2000-03-21)
This review deals with the rapid reduction of a variety of organic functionalities, including a new selective reduction of carboxylic acid, with samarium diiodide in the presence of additives under mild conditions. The single electron donor ability of samarium diiodide
Jose M Concellón et al.
Chemical Society reviews, 33(9), 599-609 (2004-12-14)
The development of methodologies for the formation of carbon-carbon double bonds could be considered one of the most important challenges in organic synthesis. To this end, beta-elimination reactions in 1,2-difunctionalised substrates have been one of the most important means of
A N Serafini
Cancer, 88(12 Suppl), 2934-2939 (2000-07-18)
In patients with bone pain due to metastatic disease, intravenous systemic radioisotope therapy may be a useful adjunct to other methods for palliating pain. Various studies have been performed utilizing a short-lived radioisotope conjugated to a tetraphosphonate (samarium 153 lexidronam)
Samarium(II)-iodide-mediated cyclizations in natural product synthesis.
David J Edmonds et al.
Chemical reviews, 104(7), 3371-3404 (2004-07-15)
Michal Szostak et al.
Chemical communications (Cambridge, England), 48(3), 330-346 (2011-09-01)
Samarium diiodide (SmI(2)) is one of the most important reductive electron transfer reagents available in the laboratory. Key to the popularity of SmI(2) is the ability of additives and co-solvents to tune the properties of the reagent. Over the last

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service