MilliporeSigma
All Photos(1)

Documents

Z405302

Best® butyl gloves

size L

Sign Into View Organizational & Contract Pricing

Select a Size

NACRES:
NB.35

material

black gloves
butyl rubber

size

L

sterility

non-sterile

feature

latex free
powder-free
reusable

packaging

pkg of 1 pair

manufacturer/tradename

Best Manufacturing 878-09

L × thickness

350 mm × 0.70 mm

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

  • Impermeable
  • Resistant to water
  • Great tactility and performance
  • Provides permeation resistance to gases and water vapors
  • Unsupported, heavy-duty, 100% butyl rubber
  • Injection molded
  • Smooth grip with rolled cuff
  • Offers flexibility and durability
  • Excellent resistance to ketones, acids, caustics, isocyanate, and gases

Legal Information

Best is a registered trademark of Showa Glove Co.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.

Visit the Document Library

Difficulty Finding Your Product Or Lot/Batch Number?

Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (example: T1503).

Example:

T1503
Product Number
-
25G
Pack Size/Quantity

Additional examples:

705578-5MG-PW

PL860-CGA/SHF-1EA

MMYOMAG-74K-13

1000309185

enter as 1.000309185)

Having trouble? Feel free to contact Technical Service for assistance.

Lot and Batch Numbers can be found on a product's label following the words 'Lot' or 'Batch'.

Aldrich Products

  • For a lot number such as TO09019TO, enter it as 09019TO (without the first two letters 'TO').

  • For a lot number with a filling-code such as 05427ES-021, enter it as 05427ES (without the filling-code '-021').

  • For a lot number with a filling-code such as STBB0728K9, enter it as STBB0728 without the filling-code 'K9'.

Not Finding What You Are Looking For?

In some cases, a COA may not be available online. If your search was unable to find the COA you can request one.

Request COA

Customers Also Viewed

Slide 1 of 2

1 of 2

2-Propanol ACS reagent, ≥99.5%

Sigma-Aldrich

190764

2-Propanol

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service