Documents
Z514470
Sigma-Aldrich
Safeskin® purple nitrile gloves
size S
Select a Size
NACRES:
NB.35
material
nitrile
purple
size
S
sterility
non-sterile
feature
disposable
powder-free
latex free
packaging
pack of 100 ea (boxes of 100)
pk of 100 ea
manufacturer/tradename
Kimberly Clark 55081
availability
not available in EU
L
9 1/2 in.
thickness
4.7 mil , palm
5.9 mil , fingertips
color
purple
General description
These unique purple gloves combine the durability of nitrile with superb comfort and fit. Powder free design reduces the risk of powder-related interference during lab work
Legal Information
Safeskin is a registered trademark of Kimberly-Clark Inc.
Certificates of Analysis (COA)
Already Own This Product?
Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.
Visit the Document Library
Difficulty Finding Your Product Or Lot/Batch Number?
Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (example: T1503).
Example:
T1503
Product Number
-
25G
Pack Size/Quantity
Additional examples:
705578-5MG-PW
PL860-CGA/SHF-1EA
MMYOMAG-74K-13
1000309185
enter as 1.000309185)
Having trouble? Feel free to contact Technical Service for assistance.
Lot and Batch Numbers can be found on a product's label following the words 'Lot' or 'Batch'.
Aldrich Products
  • For a lot number such as TO09019TO, enter it as 09019TO (without the first two letters 'TO').
  • For a lot number with a filling-code such as 05427ES-021, enter it as 05427ES (without the filling-code '-021').
  • For a lot number with a filling-code such as STBB0728K9, enter it as STBB0728 without the filling-code 'K9'.
Not Finding What You Are Looking For?
In some cases, a COA may not be available online. If your search was unable to find the COA you can request one.
Request COA