Z517178
Aldrich® modified Friedrichs condenser
jacket O.D. 37 mm, Overall H 270 mm, joint: ST/NS 24/40
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
Recommended Products
joint
joint: ST/NS24/40
Overall H
270 mm
jacket O.D.
37 mm
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Molded spiral condensing coolant tube fits closely within the jacket to force vapors along the spiral path.
Features
• Modified feed tube to reduce chance of breakage
• Additional sizes to meet specific applications
Features
• Modified feed tube to reduce chance of breakage
• Additional sizes to meet specific applications
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service