Skip to Content
MilliporeSigma

AMBH324AB855

Ambeed, Inc.

Undecanal

Synonym(s):

undecanal

Select a Size

Change View

About This Item

Linear Formula:
C11H22O1
CAS Number:
Molecular Weight:
170.3
MDL number:
Sign in to see your pricing
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C11H22O/c1-2-3-4-5-6-7-8-9-10-11-12/h11H,2-10H2,1H3

Inchi Key

KMPQYAYAQWNLME-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Inert atmosphere, room temperature

Country of Origin

CN

Physical Form

Liquid or Solid or Semi-solid or lump or cloudy liquid



Still not finding the right product?

Explore all of our products under Undecanal


hstatements

Precautionary Statements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library