Skip to Content
MilliporeSigma

AMBH2D6FD8A1

Ambeed, Inc.

Tri(pyrrolidin-1-yl)phosphine oxide

Synonym(s):

1-[di(1-pyrrolidinyl)phosphoryl]pyrrolidine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C12H24O1N3P1
CAS Number:
Molecular Weight:
257.32
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C12H24N3OP/c16-17(13-7-1-2-8-13,14-9-3-4-10-14)15-11-5-6-12-15/h1-12H2

Inchi Key

GQOGEHIVQIMJMO-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Inert atmosphere, room temperature

Country of Origin

CN

Physical Form

liquid



Still not finding the right product?


signalword

Warning

pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library