AMBH303C444F
Ambeed, Inc.
Ifenprodil tartrate
Iupac Name
4-(2-(4-benzylpiperidin-1-yl)-1-hydroxypropyl)phenol hemi((2R,3R)-2,3-dihydroxysuccinate)
Inchi Code
1S/2C21H27NO2.C4H6O6/c2*1-16(21(24)19-7-9-20(23)10-8-19)22-13-11-18(12-14-22)15-17-5-3-2-4-6-17;5-1(3(7)8)2(6)4(9)10/h2*2-10,16,18,21,23-24H,11-15H2,1H3;1-2,5-6H,(H,7,8)(H,9,10)/t;;1-,2-/m..1/s1
InChI key
DMPRDSPPYMZQBT-CEAXSRTFSA-N
storage temp.
Inert atmosphere, 2-8C
Purity
98%
Country of Origin
CN
Shipping Temperature
Normal
Physical Form
solid
Product Link
https://www.ambeed.com/products/23210-58-4.html
Synonym
4-(2-(4-Benzylpiperidin-1-yl)-1-hydroxypropyl)phenol hemi((2R,3R)-2,3-dihydroxysuccinate)
Signal Word
Warning
Hazard Statements
Precautionary statements
MSDS
https://www.ambeed.com/products/23210-58-4.html
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service