Inchi Code
1S/C5H3Cl2N/c6-4-1-5(7)3-8-2-4/h1-3H
Inchi Key
WPGHPGAUFIJVJF-UHFFFAOYSA-N
Shipping Temperature
Normal
Purity
98%
Storage temp.
Inert atmosphere, room temperature
Country of Origin
CN
Physical Form
solid
Melting Point
65-67C
Still not finding the right product?
Explore all of our products under 3,5-Dichloropyridine
hstatements
signalword
Danger
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
