Skip to Content
MilliporeSigma

AMBH303C5D48

Ambeed, Inc.

Hygromycin B

Synonym(s):

(3R,3a'S,4S,4'S,5R,6R,6'R,7'S,7a'S)-4'-(((1R,2S,3R,5S,6R)-3-amino-2,6-dihydroxy-5-(methylamino)cyclohexyl)oxy)-6-((S)-1-amino-2-hydroxyethyl)-6'-(hydroxymethyl)octahydro-4'H-spiro[pyran-2,2'-[1,3]dioxolo[4,5-c]pyran]-3,4,5,7'-tetraol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C20H37O13N3
CAS Number:
Molecular Weight:
527.53
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C20H37N3O13/c1-23-7-2-5(21)9(26)15(10(7)27)33-19-17-16(11(28)8(4-25)32-19)35-20(36-17)18(31)13(30)12(29)14(34-20)6(22)3-24/h5-19,23-31H,2-4,21-22H2,1H3/t5-,6+,7+,8-,9+,10-,11+,12-,13+,14-,15-,16+,17+,18-,19+,20?/m1/s1

Inchi Key

GRRNUXAQVGOGFE-HUCHGKBZSA-N

Shipping Temperature

Normal

Purity

950

Storage temp.

Keep in dark place, inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

solid



Still not finding the right product?

Explore all of our products under Hygromycin B



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library