Skip to Content
MilliporeSigma

AMBH97F11149

Ambeed, Inc.

D-(+)-Allose

Synonym(s):

D-allose

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C6H12O6
CAS Number:
Molecular Weight:
180.16
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C6H12O6/c7-1-3(9)5(11)6(12)4(10)2-8/h1,3-6,8-12H,2H2/t3-,4+,5-,6+/m0/s1

Inchi Key

GZCGUPFRVQAUEE-BGPJRJDNSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

solid



Still not finding the right product?

Explore all of our products under D-(+)-Allose



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library