Skip to Content
MilliporeSigma

AMBH9A6267A8

Ambeed, Inc.

Eicosadienoic acid

Synonym(s):

(11E,14E)-11,14-icosadienoic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C20H36O2
CAS Number:
Molecular Weight:
308.5
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C20H36O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20(21)22/h6-7,9-10H,2-5,8,11-19H2,1H3,(H,21,22)/b7-6+,10-9+

Inchi Key

XSXIVVZCUAHUJO-AVQMFFATSA-N

Shipping Temperature

Normal

Purity

99%

Storage temp.

Sealed in dry, store in freezer, under -20C

Country of Origin

CN

Physical Form

liquid



Still not finding the right product?

Explore all of our products under Eicosadienoic acid



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library