Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Linear Formula:
C38H36O10N6Cl2S2
CAS Number:
Molecular Weight:
871.78
MDL number:
Inchi Code
1S/C38H36Cl2N6O10S2/c39-31-11-9-29(57-31)35(49)42-18-28-20-46(38(52)56-28)26-7-3-24(4-8-26)44(36(50)30-10-12-32(40)58-30)14-16-53-21-33(47)41-17-27-19-45(37(51)55-27)25-5-1-23(2-6-25)43-13-15-54-22-34(43)48/h1-12,27-28H,13-22H2,(H,41,47)(H,42,49)/t27-,28-/m0/s1
Inchi Key
CYLLZCQTBKRWBP-NSOVKSMOSA-N
Shipping Temperature
Normal
Purity
95%
Storage temp.
Keep in dark place, inert atmosphere, 2-8C
Country of Origin
CN
Physical Form
solid
Still not finding the right product?
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.

