Skip to Content
MilliporeSigma

APO456787305

Apollo Scientific Ltd

Phleomycin

Synonym(s):

(2R,3S,4S,5R,6R)-2-(((2R,3S,4S,5S,6S)-2-(2-(6-amino-2-(3-amino-1-((2,3-diamino-3-oxopropyl)amino)-3-oxopropyl)-5-methylpyrimidine-4-carboxamido)-3-((5-((1-((2-(4-((4-guanidinobutyl)carbamoyl)-[2,4'-bithiazol]-2'-yl)ethyl)amino)-3-hydroxy-1-oxobutan-2-yl)amino)-3-hydroxy-4-methyl-5-oxopentan-2-yl)amino)-1-(1H-imidazol-4-yl)-3-oxopropoxy)-4,5-dihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-3-yl)oxy)-3,5-dihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-4-yl carbamate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
Molecular Weight:
1425.53
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C55H84N20O21S2/c1-19-32(72-45(75-43(19)58)24(11-30(57)79)67-12-23(56)44(59)85)49(89)74-34(40(25-13-63-18-68-25)94-53-42(38(83)36(81)28(14-76)93-53)95-52-39(84)41(96-55(62)91)37(82)29(15-77)92-52)50(90)69-21(3)35(80)20(2)46(86)73-33(22(4)78)48(88)65-10-7-31-70-27(17-97-31)51-71-26(16-98-51)47(87)64-8-5-6-9-66-54(60)61/h13,16-18,20-24,28-29,33-42,52-53,67,76-78,80-84H,5-12,14-15,56H2,1-4H3,(H2,57,79)(H2,59,85)(H2,62,91)(H,63,68)(H,64,87)(H,65,88)(H,69,90)(H,73,86)(H,74,89)(H2,58,72,75)(H4,60,61,66)/t20?,21?,22?,23?,24?,28-,29+,33?,34?,35?,36+,37+,38-,39-,40?,41-,42-,52+,53-/m0/s1

Inchi Key

NBLHOLNNKJBEDC-ZZLLOSFLSA-N

Country of Origin

GB



MSDS

https://store.apolloscientific.co.uk/product/phleomycin



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library