Skip to Content
MilliporeSigma

APO456787887

Apollo Scientific Ltd

Doxycycline Monohydrate

Synonym(s):

(4S,4aR,5S,5aR,6R,11aS,12aS)-4-(dimethylamino)-5,10,12a-trihydroxy-6-methyl-1,3,11,12-tetraoxo-1,2,3,4,4a,5,5a,6,11,11a,12,12a-dodecahydrotetracene-2-carboxamide hydrate

Sign In to View Organizational & Contract Pricing.

Select a Size


About This Item

CAS Number:
Molecular Weight:
462.46
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Inchi Code

1S/C22H24N2O8.H2O/c1-7-8-5-4-6-9(25)11(8)16(26)12-10(7)17(27)14-15(24(2)3)18(28)13(21(23)31)20(30)22(14,32)19(12)29;/h4-7,10,12-15,17,25,27,32H,1-3H3,(H2,23,31);1H2/t7-,10+,12-,13?,14+,15-,17-,22-;/m0./s1

Inchi Key

HLRMDPFBVRZNJZ-FBZKTRQWSA-N

Country of Origin

GB

MSDS

https://store.apolloscientific.co.uk/product/doxycycline-monohydrate


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service