Iupac Name
4-hydroxybenzene-1,2-dicarboxylic acid hydrate
Inchi Code
1S/C8H6O5.H2O/c9-4-1-2-5(7(10)11)6(3-4)8(12)13;/h1-3,9H,(H,10,11)(H,12,13);1H2
InChI key
DTZAFSPBUASGOE-UHFFFAOYSA-N
Country of Origin
GB
Product Link
https://store.apolloscientific.co.uk/product/4-hydroxyphthalic-acid-monohydrate
MSDS
https://store.apolloscientific.co.uk/product/4-hydroxyphthalic-acid-monohydrate
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service