Skip to Content
MilliporeSigma

ATEH03AB1A95

AstaTech, Inc.

OCTANE

Synonym(s):

octane

Select a Size

Change View

About This Item

Linear Formula:
C8H18
CAS Number:
Molecular Weight:
114.23
MDL number:
Sign in to see your pricing
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C8H18/c1-3-5-7-8-6-4-2/h3-8H2,1-2H3

Inchi Key

TVMXDCGIABBOFY-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

-20 C

Country of Origin

CN

Physical Form

Liquid



Still not finding the right product?

Explore all of our products under OCTANE


Signal Word

Warning



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library