Skip to Content
MilliporeSigma

ATE413072923

AstaTech, Inc.

PLATINUM(IV) OXIDE

Synonym(s):

platinum(IV) oxide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
O2Pt1
CAS Number:
Molecular Weight:
227.08
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/2O.Pt/q2*-2;+4

Inchi Key

WNVYCMIJBVIKSY-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

85%

Storage temp.

Room temperature

Country of Origin

CN

Physical Form

Yellow to Brown Solid

Melting Point

450 C(lit.)



Still not finding the right product?

Explore all of our products under PLATINUM(IV) OXIDE


hstatements

signalword

Danger



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library