Skip to Content
MilliporeSigma

ATEH11AB29F1

AstaTech, Inc.

PHOSPHOMOLYBDIC ACID XHYDRATE, =72%(BASE ON MOLYBDENUM OXIDE)

Synonym(s):

molybdic phosphoric anhydride hydrate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
H5O8P1Mo1
CAS Number:
Molecular Weight:
259.95
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/Mo.H3O4P.2H2O.2O/c;1-5(2,3)4;;;;/h;(H3,1,2,3,4);2*1H2;;/q+2;;;;;/p-2

Inchi Key

TUKGFSRCWZAYDV-UHFFFAOYSA-L

Shipping Temperature

Normal

Purity

95%

Storage temp.

2-8 C

Country of Origin

US

Physical Form

Solid





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library