Skip to Content
MilliporeSigma

ATEH98B84C8A

AstaTech, Inc.

N-(2-CARBOXYPHENYL)GLYCINE

Synonym(s):

2-[(carboxymethyl)amino]benzoic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C9H9O4N1
CAS Number:
Molecular Weight:
195.17
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C9H9NO4/c11-8(12)5-10-7-4-2-1-3-6(7)9(13)14/h1-4,10H,5H2,(H,11,12)(H,13,14)

Inchi Key

PJUXPMVQAZLJEX-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Room temperature

Country of Origin

US

Physical Form

Yellow to Brown Solid



Still not finding the right product?

Explore all of our products under N-(2-CARBOXYPHENYL)GLYCINE


signalword

Warning



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library