Skip to Content
MilliporeSigma

BL3H1F1C89D6

BLD Pharmatech

5-Bromoindole

Synonym(s):

5-bromo-1H-indole

Select a Size

Change View

About This Item

Linear Formula:
C8H6N1Br1
CAS Number:
Molecular Weight:
196.05
MDL number:
Sign in to see your pricing
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C8H6BrN/c9-7-1-2-8-6(5-7)3-4-10-8/h1-5,10H

Inchi Key

VXWVFZFZYXOBTA-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Keep in dark place, inert atmosphere, room temperature

Country of Origin

CN

Physical Form

White to yellow to brown powder or crystals

Melting Point

90-92C



Still not finding the right product?

Explore all of our products under 5-Bromoindole


hstatements

Precautionary Statements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library