Skip to Content
MilliporeSigma

BL3H9538AFA4

BLD Pharmatech

3-Chlorobenzenesulfonyl chloride

Synonym(s):

3-chlorobenzenesulfonyl chloride

Select a Size

Change View

About This Item

Linear Formula:
C6H4O2Cl2S1
CAS Number:
Molecular Weight:
211.07
MDL number:
Sign in to see your pricing
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C6H4Cl2O2S/c7-5-2-1-3-6(4-5)11(8,9)10/h1-4H

Inchi Key

OINWZUJVEXUHCC-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Inert atmosphere, room temperature

Country of Origin

CN

Physical Form

Form: Liquid - Crystal - Lump



Still not finding the right product?

Explore all of our products under 3-Chlorobenzenesulfonyl chloride


hstatements

Precautionary Statements

Signal Word

Danger

Pictograms

Corrosion


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library