Skip to Content
MilliporeSigma

BL3H160BBA6D

BLD Pharmatech

3,3-Diethoxy-1-propanol

Synonym(s):

3,3-diethoxy-1-propanol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C7H16O3
CAS Number:
Molecular Weight:
148.2
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C7H16O3/c1-3-9-7(5-6-8)10-4-2/h7-8H,3-6H2,1-2H3

Inchi Key

ASERXEZXVIJBRO-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Form: liquid Colour: colourless



Still not finding the right product?

Explore all of our products under 3,3-Diethoxy-1-propanol


hstatements

signalword

Warning

pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library