Skip to Content
MilliporeSigma

BL3H160C3BE0

BLD Pharmatech

N-(4-Nitro-3-(trifluoromethyl)phenyl)acetamide

Synonym(s):

N-[4-nitro-3-(trifluoromethyl)phenyl]acetamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C9H7O3N2F3
CAS Number:
Molecular Weight:
248.16
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C9H7F3N2O3/c1-5(15)13-6-2-3-8(14(16)17)7(4-6)9(10,11)12/h2-4H,1H3,(H,13,15)

Inchi Key

MIHJCLQINRFOLX-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Sealed in dry, room temperature

Country of Origin

CN

Physical Form

White to yellow powder or crystals



hstatements

signalword

Danger

pictograms

Corrosion


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library