Skip to Content
MilliporeSigma

BL3H1F1C8327

BLD Pharmatech

2-Bromo-5-chloropyridine

Synonym(s):

2-bromo-5-chloropyridine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C5H3N1Cl1Br1
CAS Number:
Molecular Weight:
192.44
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C5H3BrClN/c6-5-2-1-4(7)3-8-5/h1-3H

Inchi Key

BZUUVQCSPHPUQA-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Keep in dark place, sealed in dry, room temperature

Country of Origin

CN

Physical Form

Form: Crystal - Powder / Colour: White - Greyish reddish yellow



Still not finding the right product?

Explore all of our products under 2-Bromo-5-chloropyridine


signalword

Warning

pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library